missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobiphenyl, 97%

Catalog No. p-72932
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC339100010 1 g Glass bottle
AC339100050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC339100010 Supplier Thermo Scientific Chemicals Supplier No. 339100010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2113-57-7 | C12H9Br | 233.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2113-57-7
Molecular Formula C12H9Br
Molecular Weight (g/mol) 233.11
MDL Number MFCD00000082
InChI Key USYQKCQEVBFJRP-UHFFFAOYSA-N
Synonym 3-bromobiphenyl, 3-bromo-1,1'-biphenyl, 1,1'-biphenyl, 3-bromo, 3-bromo-biphenyl, m-bromobiphenyl, biphenyl, 3-bromo, 3-bromodiphenyl, unii-o7i53k56s8, ccris 5889, 3-brombiphenyl
PubChem CID 16449
IUPAC Name 1-bromo-3-phenylbenzene
SMILES BrC1=CC=CC(=C1)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Beige-Gray to Yellow
Density 1.3980g/mL
Boiling Point 300°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.6370 to 1.641
Linear Formula C6H5C6H4Br
Quantity 1 g
Specific Gravity 1.398
Formula Weight 233.11
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Bromobiphenyl, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 218-304-9
RTECSNumber DV1750000
TSCA TSCA

RUO – Research Use Only