missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. AAH58983AD
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH58983AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH58983AD Supplier Thermo Scientific Chemicals Supplier No. H58983AD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 307496-31-7 | C7H6Br2Zn | 315.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Reacts with water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-vent

Chemical Identifiers

CAS 307496-31-7
Molecular Formula C7H6Br2Zn
Molecular Weight (g/mol) 315.31
MDL Number MFCD01311434
InChI Key FBSUMSXZNCNBLE-UHFFFAOYSA-M
Synonym 3-bromobenzylzinc bromide, bromo 3-bromophenyl methyl zinc, 3-bromobenzylzinc bromide, 0.50 m in thf, 3-bromobenzylzinc bromide 0.5 m in tetrahydrofuran, 3-bromobenzylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
PubChem CID 24722602
IUPAC Name 1-bromo-3-methanidylbenzene;bromozinc(1+)
SMILES [Zn+]Br.[CH2-]C1=CC(Br)=CC=C1

Specifications

Density 1.027
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Solubility Information Reacts with water.
Formula Weight 315.31
Chemical Name or Material 3-Bromobenzylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Hazard Category H225-H260-H302-H314-H335-H336-H351
Hazard Statement GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only