missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzylmagnesium bromide, 0.25M solution in diethyl ether, AcroSeal™

Catalog No. AC431601000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC431601000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC431601000 Supplier Thermo Scientific Chemicals Supplier No. 431601000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 107549-22-4 | C7H6Br2Mg | 274.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 107549-22-4 , 60-29-7
Molecular Formula C7H6Br2Mg
Molecular Weight (g/mol) 274.24
MDL Number MFCD01319902
InChI Key OHXWEOZZKBJUTD-UHFFFAOYSA-M
Synonym 3-bromobenzylmagnesium bromide, 3-bromobenzylrnagnesium bromide, 3-bromobenzyl magnesium bromide, ohxweozzkbjutd-uhfffaoysa-m, magnesium,bromo 3-bromophenyl methyl, bromo 3-bromophenyl methyl magnesium, magnesium,1-bromo-3-methanidylbenzene,bromide, 3-bromobenzylmagnesium bromide 0.25 m in diethyl ether, 3-bromobenzylmagnesium bromide, 0.25m in diethyl ether, 3-bromobenzylmagnesium bromide solution, 0.25 m in diethyl ether, grignard reagent
PubChem CID 16217187
IUPAC Name magnesium;1-bromo-3-methanidylbenzene;bromide
SMILES Br[Mg]CC1=CC(Br)=CC=C1

Specifications

Boiling Point 34.6°C
Color Yellow
CAS Min % 5.37, 87.47
CAS Max % 12.53, 94.63
Physical Form Liquid
Quantity 100 mL
Flash Point −40°C
Formula Weight 274.24
Concentration or Composition (by Analyte or Components) 0.15 to 0.35M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.766
Density 0.7660g/mL
Chemical Name or Material 3-Bromobenzylmagnesium bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
Extremely flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only