accessibility menu, dialog, popup

UserName

CAS RN 107549-22-4

IUPAC Name: bromo[(3-bromophenyl)methyl]magnesium
Synonyms: bromo[(3-bromophenyl)methyl]magnesium
Molecular Weight (g/mol): 274.24
Molecular Formula: C7H6Br2Mg
InChi Key: OHXWEOZZKBJUTD-UHFFFAOYSA-M
SMILES: Br[Mg]CC1=CC(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results