missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzoyl Chloride 98.0+%, TCI America™

Catalog No. p-7118524
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B05575G 5 g
B055725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B05575G Supplier TCI America Supplier No. B05575G
Only null left

Chemical Identifiers

CAS 1711-09-7
Molecular Formula C7H4BrClO
Molecular Weight (g/mol) 219.462
MDL Number MFCD00000669
InChI Key PBOOZQFGWNZNQE-UHFFFAOYSA-N
Synonym m-bromobenzoyl chloride, benzoyl chloride, 3-bromo, 3-bromobenzoylchloride, benzoyl chloride, m-bromo, 3-bromobenzoic acid chloride, attercop-chm at106527, 3-brom-benzoylchlorid, 3-bromo-benzoylchlorid, 3-bromobenzoic chloride, pubchem13574
PubChem CID 74377
IUPAC Name 3-bromobenzoyl chloride
SMILES C1=CC(=CC(=C1)Br)C(=O)Cl

Specifications

Color Yellow
Boiling Point 240°C
Quantity 5 g
UN Number 3265
Formula Weight 219.46
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 3-Bromobenzoyl Chloride
EINECSNumber (3)-0062
TSCA Yes