missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzoyl chloride, 98%

Catalog No. p-7022460
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1525609 10 g
AAA1525618 50 g
AAA1525630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1525609 Supplier Thermo Scientific Chemicals Supplier No. A1525609
Only null left

CAS: 1711-09-7 | C7H4BrClO | 219.462 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1711-09-7
Molecular Formula C7H4BrClO
Molecular Weight (g/mol) 219.462
MDL Number MFCD00000669
InChI Key PBOOZQFGWNZNQE-UHFFFAOYSA-N
Synonym m-bromobenzoyl chloride, benzoyl chloride, 3-bromo, 3-bromobenzoylchloride, benzoyl chloride, m-bromo, 3-bromobenzoic acid chloride, attercop-chm at106527, 3-brom-benzoylchlorid, 3-bromo-benzoylchlorid, 3-bromobenzoic chloride, pubchem13574
PubChem CID 74377
IUPAC Name 3-bromobenzoyl chloride
SMILES C1=CC(=CC(=C1)Br)C(=O)Cl

Specifications

Density 1.666
Boiling Point 247°C to 251°C
Flash Point 107°C (224°F)
Refractive Index 1.595
Quantity 10 g
UN Number UN3265
Beilstein 1635505
Sensitivity Moisture sensitive
Formula Weight 219.47
Percent Purity 98%
Chemical Name or Material 3-Bromobenzoyl chloride
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 216-978-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only