missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzophenone, 97%

Catalog No. p-7035778
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5350503 1 g
AAH5350506 5 g
AAH5350514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5350503 Supplier Thermo Scientific Chemicals Supplier No. H5350503
Only null left

CAS: 1016-77-9 | C13H9BrO | 261.118 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1016-77-9
Molecular Formula C13H9BrO
Molecular Weight (g/mol) 261.118
MDL Number MFCD00672015
InChI Key XNUMUNIJQMSNNN-UHFFFAOYSA-N
Synonym 3-bromobenzophenone, 3-bromophenyl phenyl methanone, methanone, 3-bromophenyl phenyl, m-bromobenzophenone, 3-bromophenyl phenylmethanone, 3-bromophenyl-phenyl-methanone, 3-bromophenyl phenyl ketone, bromobenzophenone,m, 3-bromobenzoylbenzene, acmc-20an8e
PubChem CID 70548
IUPAC Name (3-bromophenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)Br

Specifications

Melting Point 74°C to 78°C
Quantity 1 g
Formula Weight 261.12
Percent Purity 97%
Chemical Name or Material 3-Bromobenzophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-808-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only