missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzohydrazide 98.0+%, TCI America™

Catalog No. p-7123380
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
B41481G 1 g
B41485G 5 g
2 options

Catalog No. B41481G

Supplier: TCI America B41481G

Only null left

Chemical Identifiers

CAS 39115-96-3
Molecular Formula C7H7BrN2O
Molecular Weight (g/mol) 215.05
MDL Number MFCD00007600
InChI Key BNAQRAZIPAHWAR-UHFFFAOYSA-N
Synonym 3-bromobenzhydrazide, m-bromobenzohydrazide, 3-bromobenzoic hydrazide, benzoic acid, 3-bromo-, hydrazide, 3-bromobenzoic acid hydrazide, 3-bromobenzoylhydrazine, benzoylhydrazine, m-bromo, 3-bromo-benzoic acid hydrazide, 3-bromobenzenecarbohydrazide, 3-bromanylbenzohydrazide
PubChem CID 520941
IUPAC Name 3-bromobenzohydrazide
SMILES C1=CC(=CC(=C1)Br)C(=O)NN

Specifications

Melting Point 157°C
Color White
Quantity 1 g
Formula Weight 215.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromobenzohydrazide