missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzo[b]thiophene 96.0+%, TCI America™

Catalog No. p-7124127
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55895G 5 g
B558925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B55895G Supplier TCI America Supplier No. B55895G
Only null left

Chemical Identifiers

CAS 7342-82-7
Molecular Formula C8H5BrS
Molecular Weight (g/mol) 213.092
MDL Number MFCD00023009
InChI Key SRWDQSRTOOMPMO-UHFFFAOYSA-N
Synonym 3-bromobenzo b thiophene, 3-bromothianaphthene, 3-bromobenzothiophene, benzo b thiophene, 3-bromo, 3-bromo-benzo b thiophene, 3-bromo benzothiophene, benzo b thiophene, 3-bromo-,, zlchem 804
PubChem CID 123250
IUPAC Name 3-bromo-1-benzothiophene
SMILES C1=CC=C2C(=C1)C(=CS2)Br

Specifications

Boiling Point 269°C
Quantity 5 g
Formula Weight 213.09
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromobenzo[b]thiophene