accessibility menu, dialog, popup

UserName

BrS

CAS RN 7342-82-7

IUPAC Name: 3-bromo-1-benzothiophene
Synonyms: 3-bromo-1-benzothiophene
Molecular Weight (g/mol): 213.09
Molecular Formula: C8H5BrS
InChi Key: SRWDQSRTOOMPMO-UHFFFAOYSA-N
SMILES: BrC1=CSC2=CC=CC=C12
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)