missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzo[b]thiophene, 95%

Catalog No. AAH3294818
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3294818 50 g
AAH3294809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3294818 Supplier Thermo Scientific Chemicals Supplier No. H3294818
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7342-82-7 | C8H5BrS | 213.092 g/mol

Antifungal activity of synthetic di (hetero) arylamines based on the benzo [b] thiophene moiety.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Antifungal activity of synthetic di (hetero) arylamines based on the benzo [b] thiophene moiety.

Solubility
Sparingly Soluble (0.14 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 7342-82-7
Molecular Formula C8H5BrS
Molecular Weight (g/mol) 213.092
MDL Number MFCD00023009
InChI Key SRWDQSRTOOMPMO-UHFFFAOYSA-N
Synonym 3-bromobenzo b thiophene, 3-bromothianaphthene, 3-bromobenzothiophene, benzo b thiophene, 3-bromo, 3-bromo-benzo b thiophene, 3-bromo benzothiophene, benzo b thiophene, 3-bromo-,, zlchem 804
PubChem CID 123250
IUPAC Name 3-bromo-1-benzothiophene
SMILES C1=CC=C2C(=C1)C(=CS2)Br

Specifications

Density 1.629
Boiling Point 72°C to 74°C (0.27 mmHg)
Refractive Index 1.668
Quantity 50 g
Solubility Information Sparingly Soluble (0.14g/L) (25°C).
Formula Weight 213.1
Percent Purity 95%
Chemical Name or Material 3-Bromobenzo[b]thiophene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Recommended Storage Ambient temperatures

RUO – Research Use Only