missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzhydrazide, 98+%

Catalog No. p-7024626
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
AAA1962806 5 g
AAA1962814 25 g
2 options

Catalog No. AAA1962806

Supplier: Thermo Scientific Chemicals A1962806

Only null left

CAS: 39115-96-3 | C7H7BrN2O | 215.05 g/mol

3-Bromobenzhydrazide is used to produce 5-(3-bromo-phenyl)-3H-[1,3,4]oxadiazole-2-thione. This reaction will need reagent potassium hydroxide and solvent ethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromobenzhydrazide is used to produce 5-(3-bromo-phenyl)-3H-[1,3,4]oxadiazole-2-thione. This reaction will need reagent potassium hydroxide and solvent ethanol.

Solubility
Slightly soluble in water (3.7 g/L) (25°C)

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 39115-96-3
Molecular Formula C7H7BrN2O
Molecular Weight (g/mol) 215.05
MDL Number MFCD00007600
InChI Key BNAQRAZIPAHWAR-UHFFFAOYSA-N
Synonym 3-bromobenzhydrazide, m-bromobenzohydrazide, 3-bromobenzoic hydrazide, benzoic acid, 3-bromo-, hydrazide, 3-bromobenzoic acid hydrazide, 3-bromobenzoylhydrazine, benzoylhydrazine, m-bromo, 3-bromo-benzoic acid hydrazide, 3-bromobenzenecarbohydrazide, 3-bromanylbenzohydrazide
PubChem CID 520941
IUPAC Name 3-bromobenzohydrazide
SMILES C1=CC(=CC(=C1)Br)C(=O)NN

Specifications

Melting Point 154°C to 157°C
Quantity 5 g
Beilstein 1945814
Solubility Information Slightly soluble in water (3.7g/L) (25°C)
Formula Weight 215.05
Percent Purity ≥98%
Chemical Name or Material 3-Bromobenzhydrazide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 254-298-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only