missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. p-7119649
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23891G 1 g
B23895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23891G Supplier TCI America Supplier No. B23891G

Chemical Identifiers

CAS 2905-24-0
Molecular Formula C6H4BrClO2S
Molecular Weight (g/mol) 255.51
MDL Number MFCD00052313
InChI Key PJGOLCXVWIYXRQ-UHFFFAOYSA-N
Synonym 3-bromobenzene-1-sulfonyl chloride, 3-bromobenzenesulphonyl chloride, m-bromobenzenesulfonyl chloride, 3-bromobenzenesulfonylchloride, m-bromobenzenesulphonyl chloride, 3-bromo-benzenesulfonyl chloride, 3-bromophenyl chlorosulfone, 3-bromophenylsulfonyl chloride, buttpark 94\57-33, benzenesulfonyl chloride, 3-bromo
PubChem CID 17943
IUPAC Name 3-bromobenzene-1-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=CC(Br)=C1

Specifications

Color Yellow
Quantity 1 g
UN Number 3265
Formula Weight 255.51
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Bromobenzenesulfonyl Chloride