missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzenesulfonamide, 97%

Catalog No. AAB2568103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2568103 1 g
AAB2568106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2568103 Supplier Thermo Scientific Chemicals Supplier No. B2568103

CAS: 89599-01-9 | C6H6BrNO2S | 236.083 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89599-01-9
Molecular Formula C6H6BrNO2S
Molecular Weight (g/mol) 236.083
MDL Number MFCD00084903
InChI Key MUBJNMWVQGHHLG-UHFFFAOYSA-N
Synonym 3-bromobenzenesulphonamide, 3-bromobenzene-1-sulfonamide, benzenesulfonamide, 3-bromo, 3-bromo-benzenesulfonamide, 3-bromobenesulfonamide, 3-bromo-benzenesulfonic acid amide, 3-bromanylbenzenesulfonamide, pubchem11759, 3-bromophenylsulfonamide, m-bromobenzenesulfonamide
PubChem CID 2734765
IUPAC Name 3-bromobenzenesulfonamide
SMILES C1=CC(=CC(=C1)Br)S(=O)(=O)N

Specifications

Melting Point 154°C to 158°C
Quantity 1 g
Formula Weight 236.09
Percent Purity 97%
Chemical Name or Material 3-Bromobenzenesulfonamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only