missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzeneboronic acid, 98+%

Catalog No. p-7046797
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1635406 5 g
AAL1635414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1635406 Supplier Thermo Scientific Chemicals Supplier No. L1635406
Only null left

CAS: 89598-96-9 | C6H6BBrO2 | 200.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89598-96-9
Molecular Formula C6H6BBrO2
Molecular Weight (g/mol) 200.83
MDL Number MFCD00239386
InChI Key AFSSVCNPDKKSRR-UHFFFAOYSA-N
Synonym 3-bromophenyl boronic acid, 3-bromobenzeneboronic acid, 3-bromophenylboronicacid, boronic acid, 3-bromophenyl, pubchem1758, 3-bromo-phenylboronic acid, acmc-209r1w, 3-bromo-benzeneboronic acid, 3-bromobenzene boronic acid
PubChem CID 2734318
IUPAC Name (3-bromophenyl)boronic acid
SMILES OB(O)C1=CC=CC(Br)=C1

Specifications

Melting Point 164°C to 168°C
Quantity 5 g
Beilstein 3127104
Formula Weight 200.83
Percent Purity ≥98%
Chemical Name or Material 3-Bromobenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only