missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzanthrone, tech. 85%

Catalog No. AAL0742222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0742222 100 g
AAL0742214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0742222 Supplier Thermo Scientific Chemicals Supplier No. L0742222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 81-96-9 | C17H9BrO | 309.162 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 81-96-9
Molecular Formula C17H9BrO
Molecular Weight (g/mol) 309.162
MDL Number MFCD00021094
InChI Key WVECFEIAZAKUNF-UHFFFAOYSA-N
Synonym 3-bromobenzanthrone, 3-brombenzanthrone, benzanthrone, 3-bromo, 7-bromomesobenzanthrone, 3-bromo-7h-benzo de anthracen-7-one, 7h-benz de anthracen-7-one, 3-bromo, 3-bromobenz d,e anthrone, 3-bromo-7h-benz de anthracen-7-one, 3-bromo-7h-benz de anthracene-7-one, unii-3dh9916fno
PubChem CID 6696
IUPAC Name 3-bromobenzo[b]phenalen-7-one
SMILES C1=CC=C2C(=C1)C3=C4C(=C(C=C3)Br)C=CC=C4C2=O

Specifications

Melting Point 164°C to 168°C
Quantity 100 g
Beilstein 1975512
Formula Weight 309.17
Percent Purity 85%
Grade Technical
Chemical Name or Material 3-Bromobenzanthrone
Hazard Category H302-H317-H331-H334
Hazard Statement GHS H Statement
H331-H302
Toxic if inhaled.
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P272-P280g-P285-P301+P312-P302+P352-P304+P340-P311-P330-P333+P313-P363-P501c
EINECSNumber 201-390-7
RTECSNumber CX5077000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only