missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzaldehyde ethylene acetal, 98+%

Catalog No. AAL1961114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1961114 25 g
AAL1961106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1961114 Supplier Thermo Scientific Chemicals Supplier No. L1961114

CAS: 17789-14-9 | C9H9BrO2 | 229.073 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17789-14-9
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00003209
InChI Key VYPYKCPWNPPBBX-UHFFFAOYSA-N
Synonym 2-3-bromophenyl-1,3-dioxolane, 3-bromobenzaldehyde ethylene acetal, 1,3-dioxolane, 2-3-bromophenyl, 1-bromo-3-1,3-dioxolan-2-yl benzene, 2-m-bromophenyl-1,3-dioxolane, m-bromobenzaldehyde ethylene acetal, pubchem3730, 2-3-bromophenyl dioxolane, acmc-1c8q7, ksc496m7r
PubChem CID 87306
IUPAC Name 2-(3-bromophenyl)-1,3-dioxolane
SMILES C1COC(O1)C2=CC(=CC=C2)Br

Specifications

Density 1.514
Boiling Point 132°C to 133°C (8 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.563
Quantity 25 g
Beilstein 1344030
Formula Weight 229.07
Percent Purity ≥98%
Chemical Name or Material 3-Bromobenzaldehyde ethylene acetal
EINECSNumber 241-766-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only