missing translation for 'onlineSavingsMsg'
Learn More

3-Bromoanisole 98.0+%, TCI America™

Catalog No. B054525G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B054525G 25 g
B0545250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B054525G Supplier TCI America Supplier No. B054525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2398-37-0
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.04
MDL Number MFCD00000081
InChI Key PLDWAJLZAAHOGG-UHFFFAOYSA-N
Synonym 3-bromoanisole, m-bromoanisole, 3-methoxybromobenzene, 3-bromo anisole, m-bromomethoxybenzene, benzene, 1-bromo-3-methoxy, anisole, m-bromo, 3-methoxy-1-bromobenzene, m-methoxybromobenzene, meta-bromoanisole
PubChem CID 16971
IUPAC Name 1-bromo-3-methoxybenzene
SMILES COC1=CC=CC(Br)=C1

Specifications

Color Yellow
Boiling Point 211°C
Quantity 25 g
Formula Weight 187.04
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromoanisole