missing translation for 'onlineSavingsMsg'
Learn More

3-Bromoanisole, 98+%

Catalog No. p-7022727
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1562514 25 g
AAA1562522 100 g
AAA1562536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1562514 Supplier Thermo Scientific Chemicals Supplier No. A1562514

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2398-37-0 | C7H7BrO | 187.04 g/mol

3-Bromoanisole used in chemical reaction as intermediates to obtain target materials such as dyes, pharmaceuticals, perfumes, photoinitiators and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromoanisole used in chemical reaction as intermediates to obtain target materials such as dyes, pharmaceuticals, perfumes, photoinitiators and agrochemicals.

Solubility
Miscible in methanol, benzene, hexane, toluene and dichloromethane. Practically insoluble in water.

Notes
Store at room temperature. Store in cool dry place in tightly closed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 2398-37-0
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.04
MDL Number MFCD00000081
InChI Key PLDWAJLZAAHOGG-UHFFFAOYSA-N
Synonym 3-bromoanisole, m-bromoanisole, 3-methoxybromobenzene, 3-bromo anisole, m-bromomethoxybenzene, benzene, 1-bromo-3-methoxy, anisole, m-bromo, 3-methoxy-1-bromobenzene, m-methoxybromobenzene, meta-bromoanisole
PubChem CID 16971
IUPAC Name 1-bromo-3-methoxybenzene
SMILES COC1=CC=CC(Br)=C1

Specifications

Melting Point ∼2°C
Density 1.489
Boiling Point 210°C to 211°C
Flash Point 93°C (199°F)
Refractive Index 1.564
Quantity 25 g
Beilstein 1524687
Solubility Information Miscible in methanol,benzene,hexane,toluene and dichloromethane. Practically insoluble in water.
Formula Weight 187.04
Percent Purity ≥98%
Chemical Name or Material 3-Bromoanisole
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 219-264-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only