missing translation for 'onlineSavingsMsg'
Learn More

3-Bromoaniline 98.0+%, TCI America™

Catalog No. B054225G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B054225G 25 g
B0542100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B054225G Supplier TCI America Supplier No. B054225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 591-19-5
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.03
MDL Number MFCD00007757
InChI Key DHYHYLGCQVVLOQ-UHFFFAOYSA-N
Synonym m-bromoaniline, benzenamine, 3-bromo, 3-bromo aniline, m-aminobromobenzene, aniline, m-bromo, 3-bromobenzenamine, 3-bromophenylamine, bromoaniline 3-, 3-bromo-benzenamine, 3-bromo-phenylamine
PubChem CID 11562
IUPAC Name 3-bromoaniline
SMILES NC1=CC=CC(Br)=C1

Specifications

Color Yellow
Boiling Point 251°C
Quantity 25 g
Formula Weight 172.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromoaniline
EINECSNumber (3)-0221
RTECSNumber CX9855300
TSCA Yes