missing translation for 'onlineSavingsMsg'
Learn More

3-Bromoaniline, 98%

Catalog No. AAA1366536
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1366536 500 g
AAA1366514 25 g
AAA1366522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1366536 Supplier Thermo Scientific Chemicals Supplier No. A1366536
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 591-19-5 | C6H6BrN | 172.03 g/mol

3-Bromoaniline is used in the synthesis of amino substituted quinazoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromoaniline is used in the synthesis of amino substituted quinazoline.

Solubility
Soluble in water (0.97 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 591-19-5
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.03
MDL Number MFCD00007757
InChI Key DHYHYLGCQVVLOQ-UHFFFAOYSA-N
Synonym m-bromoaniline, benzenamine, 3-bromo, 3-bromo aniline, m-aminobromobenzene, aniline, m-bromo, 3-bromobenzenamine, 3-bromophenylamine, bromoaniline 3-, 3-bromo-benzenamine, 3-bromo-phenylamine
PubChem CID 11562
IUPAC Name 3-bromoaniline
SMILES NC1=CC=CC(Br)=C1

Specifications

Melting Point 16°C
Density 1.59
Boiling Point 251°C
Flash Point >110°C (230°F)
Odor Characteristic
Assay Percent Range 98%
Refractive Index 1.625
Quantity 500 g
UN Number UN2810
Beilstein 742028
Solubility Information Soluble in water (0.97g/L at 20°C).
Formula Weight 172.03
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Bromoaniline
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H311-H373-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 209-704-4
RTECSNumber CX9855300
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only