missing translation for 'onlineSavingsMsg'
Learn More

3-Bromoadamantane-1-carboxylic acid, 97%

Catalog No. aah6058003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6058003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6058003 Supplier Thermo Scientific Chemicals Supplier No. H6058003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 21816-08-0 | C11H15BrO2 | 259.143 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21816-08-0
Molecular Formula C11H15BrO2
Molecular Weight (g/mol) 259.143
MDL Number MFCD00167820
InChI Key DJUDQBVINJIMFO-UHFFFAOYSA-N
Synonym 1-bromo-3-adamantanecarboxylic acid, 3-bromo-1-adamantanecarboxylic acid, tricyclo 3.3.1.13,7 decane-1-carboxylic acid, 3-bromo, 3-bromo-adamantane-1-carboxylic acid, 3-adamantanecarboxylic acid, 1-bromo, 3-bromoadamantanecarboxylic acid, 3-bromotricyclo 3.3.1.1∼3,7∼ decane-1-carboxylic acid, acmc-1cnbw, maybridge1_005590, 5s,7r-3-bromoadamantane-1-carboxylate
PubChem CID 30818
IUPAC Name 3-bromoadamantane-1-carboxylic acid
SMILES C1C2CC3(CC1CC(C2)(C3)Br)C(=O)O

Specifications

Melting Point 146°C to 150°C
Quantity 1 g
Formula Weight 259.14
Percent Purity 97%
Chemical Name or Material 3-Bromoadamantane-1-carboxylic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber AU4452500
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only