missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Bromoacetyl)coumarin 95.0+%, TCI America™
$52.19 - $143.54
Chemical Identifiers
| CAS | 29310-88-1 |
|---|---|
| Molecular Formula | C11H7BrO3 |
| Molecular Weight (g/mol) | 267.08 |
| MDL Number | MFCD00488331 |
| InChI Key | NTYOLVNSXVYRTJ-UHFFFAOYSA-N |
| Synonym | 3-bromoacetyl coumarin, 3-2-bromoacetyl-2h-chromen-2-one, 3-2-bromoacetyl chromen-2-one, 3-bromoacetylcoumarin, 3-bromoacetyl-2h-chromen-2-one, 2h-1-benzopyran-2-one, 3-bromoacetyl, pubchem8670, rarechem ab ka k003, 3-2-bromoacetyl-coumarin |
| PubChem CID | 2063461 |
| IUPAC Name | 3-(2-bromoacetyl)-2H-chromen-2-one |
| SMILES | BrCC(=O)C1=CC2=CC=CC=C2OC1=O |
Chemical Identifiers
| 29310-88-1 | |
| 267.08 | |
| NTYOLVNSXVYRTJ-UHFFFAOYSA-N | |
| 2063461 | |
| BrCC(=O)C1=CC2=CC=CC=C2OC1=O |
| C11H7BrO3 | |
| MFCD00488331 | |
| 3-bromoacetyl coumarin, 3-2-bromoacetyl-2h-chromen-2-one, 3-2-bromoacetyl chromen-2-one, 3-bromoacetylcoumarin, 3-bromoacetyl-2h-chromen-2-one, 2h-1-benzopyran-2-one, 3-bromoacetyl, pubchem8670, rarechem ab ka k003, 3-2-bromoacetyl-coumarin | |
| 3-(2-bromoacetyl)-2H-chromen-2-one |
Specifications
| 29310-88-1 | |
| White-Yellow | |
| MFCD00488331 | |
| 3-bromoacetyl coumarin, 3-2-bromoacetyl-2h-chromen-2-one, 3-2-bromoacetyl chromen-2-one, 3-bromoacetylcoumarin, 3-bromoacetyl-2h-chromen-2-one, 2h-1-benzopyran-2-one, 3-bromoacetyl, pubchem8670, rarechem ab ka k003, 3-2-bromoacetyl-coumarin | |
| BrCC(=O)C1=CC2=CC=CC=C2OC1=O | |
| 267.08 | |
| 267.08 | |
| Crystalline Powder |
| 165°C | |
| C11H7BrO3 | |
| 1 g | |
| NTYOLVNSXVYRTJ-UHFFFAOYSA-N | |
| 3-(2-bromoacetyl)-2H-chromen-2-one | |
| 2063461 | |
| ≥95.0% (GC) | |
| 3-(Bromoacetyl)coumarin |
Safety and Handling
TSCA : No