CAS RN 29310-88-1
CAS RN 29310-88-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-(Bromoacetyl)coumarin 95.0+%, TCI America™
CAS: 29310-88-1 Molecular Formula: C11H7BrO3 Molecular Weight (g/mol): 267.08 MDL Number: MFCD00488331 InChI Key: NTYOLVNSXVYRTJ-UHFFFAOYSA-N Synonym: 3-bromoacetyl coumarin,3-2-bromoacetyl-2h-chromen-2-one,3-2-bromoacetyl chromen-2-one,3-bromoacetylcoumarin,3-bromoacetyl-2h-chromen-2-one,2h-1-benzopyran-2-one, 3-bromoacetyl,pubchem8670,rarechem ab ka k003,3-2-bromoacetyl-coumarin PubChem CID: 2063461 IUPAC Name: 3-(2-bromoacetyl)-2H-chromen-2-one SMILES: BrCC(=O)C1=CC2=CC=CC=C2OC1=O