missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Bromoacetyl)coumarin 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 165°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 267.08 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-(Bromoacetyl)coumarin |
Safety and Handling
| TSCA | No |