missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-p-anisaldehyde 98.0+%, TCI America™

Catalog No. p-7119264
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
B17905G 5 g
B179025G 25 g
2 options

Catalog No. B17905G

Supplier: TCI America B17905G

Only null left

Chemical Identifiers

CAS 34841-06-0
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00016599
InChI Key QMPNFQLVIGPNEI-UHFFFAOYSA-N
Synonym 3-bromo-p-anisaldehyde, benzaldehyde, 3-bromo-4-methoxy, 3-bromo-4-methoxy-benzaldehyde, 3-bromoanisaldehyde, 5-bromoanisaldehyde, 3-bromo-anisaldehyde, 5-bromo anisaldehyde, pubchem2643, acmc-209iat, 2-bromo-4-formylanisole
PubChem CID 98662
IUPAC Name 3-bromo-4-methoxybenzaldehyde
SMILES COC1=CC=C(C=O)C=C1Br

Specifications

Melting Point 50°C
Color White-Yellow
Boiling Point 162°C
Quantity 5 g
Formula Weight 215.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-p-anisaldehyde