missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-L-phenylalanine, 95%

CAS: 82311-69-1 | C9H10BrNO2 | 244.09 g/mol

Supplier:  Thermo Scientific Chemicals H5211403

 View more versions of this product

Catalog No. AAH5211403


Only null left

Description

Description

It is applied in the facile removal of leader peptides from lanthipeptides by incorporation of a hydroxy acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in the facile removal of leader peptides from lanthipeptides by incorporation of a hydroxy acid.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

3-Bromo-L-phenylalanine
C9H10BrNO2
3-bromo-l-phenylalanine, s-2-amino-3-3-bromophenyl propanoic acid, l-3-bromophenylalanine, h-phe 3-br-oh, 2s-2-amino-3-3-bromophenyl propanoic acid, l-3-bromophe, l-3-br-phe-oh, l-bromophenyl alanine, s-3-bromophenylalanine, 3-br-l-phe-oh
GDMOHOYNMWWBAU-UHFFFAOYNA-N
(2S)-2-amino-3-(3-bromophenyl)propanoic acid
244.09
244.09
82311-69-1
MFCD06659110
Slightly soluble in water.
NC(CC1=CC(Br)=CC=C1)C(O)=O
1 g
2762259
95%

Chemical Identifiers

82311-69-1
244.09
GDMOHOYNMWWBAU-UHFFFAOYNA-N
2762259
NC(CC1=CC(Br)=CC=C1)C(O)=O
C9H10BrNO2
MFCD06659110
3-bromo-l-phenylalanine, s-2-amino-3-3-bromophenyl propanoic acid, l-3-bromophenylalanine, h-phe 3-br-oh, 2s-2-amino-3-3-bromophenyl propanoic acid, l-3-bromophe, l-3-br-phe-oh, l-bromophenyl alanine, s-3-bromophenylalanine, 3-br-l-phe-oh
(2S)-2-amino-3-(3-bromophenyl)propanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only