accessibility menu, dialog, popup

UserName

CAS RN 82311-69-1

IUPAC Name: 2-amino-3-(3-bromophenyl)propanoic acid
Synonyms: 2-amino-3-(3-bromophenyl)propanoic acid
Molecular Weight (g/mol): 244.09
Molecular Formula: C9H10BrNO2
InChi Key: GDMOHOYNMWWBAU-UHFFFAOYNA-N
SMILES: NC(CC1=CC(Br)=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)