missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-L-phenylalanine, 95%

Catalog No. AAH52114MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52114MD 250 mg
AAH5211403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52114MD Supplier Thermo Scientific Chemicals Supplier No. H52114MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 82311-69-1 | C9H10BrNO2 | 244.09 g/mol

It is applied in the facile removal of leader peptides from lanthipeptides by incorporation of a hydroxy acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in the facile removal of leader peptides from lanthipeptides by incorporation of a hydroxy acid.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 82311-69-1
Molecular Formula C9H10BrNO2
Molecular Weight (g/mol) 244.09
MDL Number MFCD06659110
InChI Key GDMOHOYNMWWBAU-UHFFFAOYNA-N
Synonym 3-bromo-l-phenylalanine, s-2-amino-3-3-bromophenyl propanoic acid, l-3-bromophenylalanine, h-phe 3-br-oh, 2s-2-amino-3-3-bromophenyl propanoic acid, l-3-bromophe, l-3-br-phe-oh, l-bromophenyl alanine, s-3-bromophenylalanine, 3-br-l-phe-oh
PubChem CID 2762259
IUPAC Name (2S)-2-amino-3-(3-bromophenyl)propanoic acid
SMILES NC(CC1=CC(Br)=CC=C1)C(O)=O

Specifications

Solubility Information Slightly soluble in water.
Quantity 250 mg
Formula Weight 244.09
Percent Purity 95%
Chemical Name or Material 3-Bromo-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only