missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-9,9-diphenyl-9H-fluorene 98.0+%, TCI America™

Catalog No. b56181g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B56181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B56181G Supplier TCI America Supplier No. B56181G

Chemical Identifiers

CAS 1547491-70-2
Molecular Formula C25H17Br
Molecular Weight (g/mol) 397.315
MDL Number MFCD24849694
InChI Key AXVLLFWWMWDULG-UHFFFAOYSA-N
PubChem CID 90026835
IUPAC Name 3-bromo-9,9-diphenylfluorene
SMILES C1=CC=C(C=C1)C2(C3=C(C=C(C=C3)Br)C4=CC=CC=C42)C5=CC=CC=C5

Specifications

Melting Point 223°C
Color White-Yellow
Quantity 1 g
Formula Weight 397.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-9,9-diphenyl-9H-fluorene