missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-9,9-dimethyl-9H-fluorene 98.0+%, TCI America™

Catalog No. B5436200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5436200MG 200 mg
B54361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5436200MG Supplier TCI America Supplier No. B5436200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1190360-23-6
Molecular Formula C15H13Br
Molecular Weight (g/mol) 273.173
MDL Number MFCD28955974
InChI Key VECLPBKDHAALGQ-UHFFFAOYSA-N
PubChem CID 59629661
IUPAC Name 3-bromo-9,9-dimethylfluorene
SMILES CC1(C2=C(C=C(C=C2)Br)C3=CC=CC=C31)C

Specifications

Melting Point 63°C
Color White-Yellow
Quantity 200 mg
Formula Weight 273.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-9,9-dimethyl-9H-fluorene