missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-6-chloroimidazo[1,2-b]pyridazine, 95%

Catalog No. AAH6653106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6653106 5 g
AAH6653103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6653106 Supplier Thermo Scientific Chemicals Supplier No. H6653106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13526-66-4 | C6H3BrClN3 | 232.465 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13526-66-4
Molecular Formula C6H3BrClN3
Molecular Weight (g/mol) 232.465
MDL Number MFCD09027274
InChI Key PFHPKMPWBFJZEY-UHFFFAOYSA-N
Synonym 3-bromo-6-chloroimidazo 1,2-b pyridazine, 6-chloro-3-bromoimidazo 1,2-a pyridazine, imidazo 1,2-b pyridazine, 3-bromo-6-chloro, 3-bromo-6-chloro-imidazo 1,2-b pyridazine, librarion l936, pubchem23320, ksc496o7t, 3-brom-6-chlorimidazo 1,2-b pyridazin, 6-chloro-3-bromoimidazo 1,2-b pyridazine
PubChem CID 12872323
IUPAC Name 3-bromo-6-chloroimidazo[1,2-b]pyridazine
SMILES C1=CC(=NN2C1=NC=C2Br)Cl

Specifications

Quantity 5 g
Beilstein 513113
Formula Weight 232.47
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Bromo-6-chloroimidazo[1,2-b]pyridazine
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only