accessibility menu, dialog, popup

UserName

CAS RN 13526-66-4

IUPAC Name: 3-bromo-6-chloroimidazo[1,2-b]pyridazine
Synonyms: 3-bromo-6-chloroimidazo[1,2-b]pyridazine
Molecular Weight (g/mol): 232.47
Molecular Formula: C6H3BrClN3
InChi Key: PFHPKMPWBFJZEY-UHFFFAOYSA-N
SMILES: ClC1=NN2C(Br)=CN=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results