missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-(trifluoromethyl)benzenesulfonyl chloride, 97%

Catalog No. AAL1971103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1971103 1 g
AAL1971106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1971103 Supplier Thermo Scientific Chemicals Supplier No. L1971103

CAS: 351003-46-8 | C7H3BrClF3O2S | 323.508 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 351003-46-8
Molecular Formula C7H3BrClF3O2S
Molecular Weight (g/mol) 323.508
MDL Number MFCD03094393
InChI Key YBUJCZFWJSUTSN-UHFFFAOYSA-N
Synonym 3-bromo-5-trifluoromethyl benzenesulfonyl chloride, 3-bromo-5-trifluoromethyl benzene-1-sulfonyl chloride, 3-bromo-5-trifluoromethyl benzenesulphonylchloride, 3-bromo-5-trifluoromethyl benzene sulphonyl chloride, 5-bromo-3-trifluoromethyl phenyl chlorosulfone, 3-bromo-5-trifluoromethylphenylsulphonyl chloride, 3-bromo-5-trifluoromethyl benzenesulphonyl chloride, benzenesulfonylchloride, 3-bromo-5-trifluoromethyl, 3-bromo-5-trifluoromethylphenylsulfonyl chloride, 3-bromo-5-trifluoromethyl benzenesulfonylchloride
PubChem CID 2778610
IUPAC Name 3-bromo-5-(trifluoromethyl)benzenesulfonyl chloride
SMILES C1=C(C=C(C=C1S(=O)(=O)Cl)Br)C(F)(F)F

Specifications

Density 1.835
Boiling Point 217°C to 218°C
Flash Point 66°C (150°F)
Refractive Index 1.524
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 323.52
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-(trifluoromethyl)benzenesulfonyl chloride
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only