missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-(trifluoromethyl)benzaldehyde 98.0+%, TCI America™

Catalog No. B49741G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49741G 1 g
B49745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49741G Supplier TCI America Supplier No. B49741G

Chemical Identifiers

CAS 477535-41-4
Molecular Formula C8H4BrF3O
Molecular Weight (g/mol) 253.02
MDL Number MFCD08059506
InChI Key PCRLZGCXLNNMFL-UHFFFAOYSA-N
Synonym 3-bromo-5-trifluoromethyl benzaldehyde, benzaldehyde, 3-bromo-5-trifluoromethyl, 3-trifluoromethyl-5-bromobenzaldehyde, 3-bromo-5-trifluoromethyl-benzaldehyde, pubchem16131, acmc-209kad, ksc496e1r, 3-bromo-5-formylbenzotrifluoride, 5-bromo-3-trifluoromethyl benzaldehyde
PubChem CID 16115438
IUPAC Name 3-bromo-5-(trifluoromethyl)benzaldehyde
SMILES FC(F)(F)C1=CC(Br)=CC(C=O)=C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 253.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromo-5-(trifluoromethyl)benzaldehyde