missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-nitrosalicylaldehyde 95.0+%, TCI America™

Catalog No. B31981G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31981G 1 g
B31985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31981G Supplier TCI America Supplier No. B31981G

Chemical Identifiers

CAS 16789-84-7
Molecular Formula C7H4BrNO4
Molecular Weight (g/mol) 246.02
MDL Number MFCD00051833
InChI Key BESBCGANGAEHPM-UHFFFAOYSA-N
Synonym 3-bromo-2-hydroxy-5-nitrobenzaldehyde, 3-bromo-5-nitrosalicylaldehyde, 3-bromo-2-hydroxy-5-nitrobenzenecarbaldehyde, 2-hydroxy-3-bromo-5-nitrobenzaldehyde, 3-bromo-2-hydroxy-5-nitro-benzaldehyde, acmc-1cfaz, 3-bromo-5-nitro-salicylaldehyde, benzaldehyde,3-bromo-2-hydroxy-5-nitro, 3-bromanyl-5-nitro-2-oxidanyl-benzaldehyde
PubChem CID 519307
IUPAC Name 3-bromo-2-hydroxy-5-nitrobenzaldehyde
SMILES OC1=C(Br)C=C(C=C1C=O)[N+]([O-])=O

Specifications

Melting Point 149°C
Color Yellow
Quantity 1 g
UN Number 2811
Formula Weight 246.02
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-5-nitrosalicylaldehyde