missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-nitrosalicylaldehyde, 98%

Catalog No. AAL0707103
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0707103 1 g
AAL0707106 5 g
AAL0707114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0707103 Supplier Thermo Scientific Chemicals Supplier No. L0707103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 16789-84-7 | C7H4BrNO4 | 246.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16789-84-7
Molecular Formula C7H4BrNO4
Molecular Weight (g/mol) 246.02
MDL Number MFCD00051833
InChI Key BESBCGANGAEHPM-UHFFFAOYSA-N
Synonym 3-bromo-2-hydroxy-5-nitrobenzaldehyde, 3-bromo-5-nitrosalicylaldehyde, 3-bromo-2-hydroxy-5-nitrobenzenecarbaldehyde, 2-hydroxy-3-bromo-5-nitrobenzaldehyde, 3-bromo-2-hydroxy-5-nitro-benzaldehyde, acmc-1cfaz, 3-bromo-5-nitro-salicylaldehyde, benzaldehyde,3-bromo-2-hydroxy-5-nitro, 3-bromanyl-5-nitro-2-oxidanyl-benzaldehyde
PubChem CID 519307
IUPAC Name 3-bromo-2-hydroxy-5-nitrobenzaldehyde
SMILES OC1=C(Br)C=C(C=C1C=O)[N+]([O-])=O

Specifications

Melting Point 147°C to 151°C
Quantity 1 g
Beilstein 1213613
Sensitivity Air Sensitive
Formula Weight 246.02
Percent Purity 98%
Chemical Name or Material 3-Bromo-5-nitrosalicylaldehyde
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only