missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-5-methoxytoluene 98.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 122°C |
| Quantity | 5 g |
| Formula Weight | 201.06 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-Bromo-5-methoxytoluene |
Safety and Handling
| TSCA | No |