missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-methoxybenzoic acid, 98%

Catalog No. p-7031525
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3263503 1 g
AAH3263506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3263503 Supplier Thermo Scientific Chemicals Supplier No. H3263503

CAS: 157893-14-6 | C8H7BrO3 | 231.045 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 157893-14-6
Molecular Formula C8H7BrO3
Molecular Weight (g/mol) 231.045
MDL Number MFCD00266858
InChI Key DMXJBCHYVUGXEH-UHFFFAOYSA-N
Synonym 3-bromo-5-carboxyanisole, 3-bromo-5-methoxybenzoicacid, 3-bromo-5-methoxy-benzoic acid, benzoic acid, 3-bromo-5-methoxy, 5-bromo-3-methoxybenzoic acid, pubchem16435, acmc-1bx98, ksc529k1f, 3-bromo-5-methyloxy benzoic acid
PubChem CID 4060048
IUPAC Name 3-bromo-5-methoxybenzoic acid
SMILES COC1=CC(=CC(=C1)C(=O)O)Br

Specifications

Quantity 1 g
Formula Weight 231.05
Percent Purity 98%
Chemical Name or Material 3-Bromo-5-methoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only