missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-iodopyridine 98.0+%, TCI America™

Catalog No. p-7123484
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43461G 1 g
B43465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43461G Supplier TCI America Supplier No. B43461G
Only null left

Chemical Identifiers

CAS 233770-01-9
Molecular Formula C5H3BrIN
Molecular Weight (g/mol) 283.894
MDL Number MFCD03086019
InChI Key AOOZLVWDZUPEHT-UHFFFAOYSA-N
PubChem CID 820409
IUPAC Name 3-bromo-5-iodopyridine
SMILES C1=C(C=NC=C1I)Br

Specifications

Melting Point 133°C
Color White-Yellow
Quantity 1 g
Synonym 3-bromo-5-iodo-pyridine, 5-bromo-3-iodopyridine, pyridine, 3-bromo-5-iodo, 3-bromo-5-iodpyridine, 3-iodo-5-bromopyridine, pubchem8080, acmc-1cdm9, pyridine,3-bromo-5-iodo, 3-bromo-5-iodopyridine, abbypharma ap-13-5787
Formula Weight 283.89
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-5-iodopyridine