missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-5-hydroxybenzoic Acid 98.0+%, TCI America™
$91.33 - $272.16
Chemical Identifiers
| CAS | 140472-69-1 |
|---|---|
| Molecular Formula | C7H5BrO3 |
| Molecular Weight (g/mol) | 217.018 |
| MDL Number | MFCD06797980 |
| InChI Key | WGIBEMRBLBGETQ-UHFFFAOYSA-N |
| PubChem CID | 15735511 |
| IUPAC Name | 3-bromo-5-hydroxybenzoic acid |
| SMILES | C1=C(C=C(C=C1O)Br)C(=O)O |
Chemical Identifiers
| 140472-69-1 | |
| 217.018 | |
| WGIBEMRBLBGETQ-UHFFFAOYSA-N | |
| 3-bromo-5-hydroxybenzoic acid |
| C7H5BrO3 | |
| MFCD06797980 | |
| 15735511 | |
| C1=C(C=C(C=C1O)Br)C(=O)O |
Specifications
| 140472-69-1 | |
| C7H5BrO3 | |
| 1 g | |
| WGIBEMRBLBGETQ-UHFFFAOYSA-N | |
| 3-bromo-5-hydroxybenzoic acid | |
| 15735511 | |
| ≥98.0% (GC,T) | |
| 3-Bromo-5-hydroxybenzoic Acid |
| 239°C | |
| MFCD06797980 | |
| 3077 | |
| C1=C(C=C(C=C1O)Br)C(=O)O | |
| 217.018 | |
| 217.02 | |
| Crystalline Powder |
Safety and Handling
TSCA : No