missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-hydroxybenzoic Acid 98.0+%, TCI America™

Catalog No. B47405G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47405G 5 g
B47401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47405G Supplier TCI America Supplier No. B47405G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 140472-69-1
Molecular Formula C7H5BrO3
Molecular Weight (g/mol) 217.018
MDL Number MFCD06797980
InChI Key WGIBEMRBLBGETQ-UHFFFAOYSA-N
PubChem CID 15735511
IUPAC Name 3-bromo-5-hydroxybenzoic acid
SMILES C1=C(C=C(C=C1O)Br)C(=O)O

Specifications

Melting Point 239°C
Quantity 5 g
UN Number 3077
Formula Weight 217.02
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-5-hydroxybenzoic Acid