missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-5-hydroxybenzoic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 239°C |
| Quantity | 1 g |
| UN Number | 3077 |
| Formula Weight | 217.02 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Bromo-5-hydroxybenzoic Acid |
Safety and Handling
| TSCA | No |