missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-fluoropyridine 98.0+%, TCI America™

Catalog No. p-7122831
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32441G 1 g
B32445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32441G Supplier TCI America Supplier No. B32441G
Only null left

Chemical Identifiers

CAS 407-20-5
Molecular Formula C5H3BrFN
Molecular Weight (g/mol) 175.99
MDL Number MFCD04112555
InChI Key HNNNBQRRIHKFLI-UHFFFAOYSA-N
Synonym 5-bromo-3-fluoropyridine, 3-bromo-5-fluoro-pyridine, 3-brom-5-fluorpyridin, 3-bromo-5-fluoro pyridine, pyridine, 3-bromo-5-fluoro, 3-bromo-5-fluorpyridine, 5-bromo-3-fluoride pyridine, 3-bromine-5-fluoride pyridine, pubchem3964, 3-bromo-5-fluoropyridin
PubChem CID 820206
IUPAC Name 3-bromo-5-fluoropyridine
SMILES FC1=CN=CC(Br)=C1

Specifications

Color White-Yellow
Boiling Point 78°C
Quantity 1 g
Formula Weight 175.99
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 3-Bromo-5-fluoropyridine