missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-fluoropyridine, 97%

Catalog No. p-7028443
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2576603 1 g
AAH2576606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2576603 Supplier Thermo Scientific Chemicals Supplier No. H2576603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 407-20-5 | C5H3BrFN | 175.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 407-20-5
Molecular Formula C5H3BrFN
Molecular Weight (g/mol) 175.99
MDL Number MFCD04112555
InChI Key HNNNBQRRIHKFLI-UHFFFAOYSA-N
Synonym 5-bromo-3-fluoropyridine, 3-bromo-5-fluoro-pyridine, 3-brom-5-fluorpyridin, 3-bromo-5-fluoro pyridine, pyridine, 3-bromo-5-fluoro, 3-bromo-5-fluorpyridine, 5-bromo-3-fluoride pyridine, 3-bromine-5-fluoride pyridine, pubchem3964, 3-bromo-5-fluoropyridin
PubChem CID 820206
IUPAC Name 3-bromo-5-fluoropyridine
SMILES FC1=CN=CC(Br)=C1

Specifications

Melting Point 24°C to 28°C
Flash Point 64°C (148°F)
Odor Characteristic
Quantity 1 g
UN Number UN2811
Formula Weight 175.99
Percent Purity 99%
Chemical Name or Material 3-Bromo-5-fluoropyridine
Hazard Category H302-H315-H318-H335-H500
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only