missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-5-fluorobenzonitrile 98.0+%, TCI America™
$37.29 - $139.81
Chemical Identifiers
| CAS | 179898-34-1 |
|---|---|
| Molecular Formula | C7H3BrFN |
| Molecular Weight (g/mol) | 200.01 |
| MDL Number | MFCD04038227 |
| InChI Key | IADLVSLZPQYXIF-UHFFFAOYSA-N |
| Synonym | 3-fluoro-5-bromobenzonitrile, benzonitrile, 3-bromo-5-fluoro, 5-bromo-3-fluorobenzenecarbonitrile, 3-cyano-5-fluorobromobenzene, 3-bromo 5-fluorobenzonitrile, 3-bromo-5-fluoro-benzonitrile, 3-bromo-5-fluorobenzenecarbonitrile, pubchem3220, acmc-209yem, 3-fluoro-5-bromobenzonitile |
| PubChem CID | 2783330 |
| IUPAC Name | 3-bromo-5-fluorobenzonitrile |
| SMILES | FC1=CC(Br)=CC(=C1)C#N |
Chemical Identifiers
| 179898-34-1 | |
| 200.01 | |
| IADLVSLZPQYXIF-UHFFFAOYSA-N | |
| 2783330 | |
| FC1=CC(Br)=CC(=C1)C#N |
| C7H3BrFN | |
| MFCD04038227 | |
| 3-fluoro-5-bromobenzonitrile, benzonitrile, 3-bromo-5-fluoro, 5-bromo-3-fluorobenzenecarbonitrile, 3-cyano-5-fluorobromobenzene, 3-bromo 5-fluorobenzonitrile, 3-bromo-5-fluoro-benzonitrile, 3-bromo-5-fluorobenzenecarbonitrile, pubchem3220, acmc-209yem, 3-fluoro-5-bromobenzonitile | |
| 3-bromo-5-fluorobenzonitrile |
Specifications
| 179898-34-1 | |
| White | |
| MFCD04038227 | |
| 3439 | |
| IADLVSLZPQYXIF-UHFFFAOYSA-N | |
| 3-bromo-5-fluorobenzonitrile | |
| 2783330 | |
| ≥98.0% (GC) | |
| 3-Bromo-5-fluorobenzonitrile |
| 46°C | |
| C7H3BrFN | |
| 1 g | |
| 3-fluoro-5-bromobenzonitrile, benzonitrile, 3-bromo-5-fluoro, 5-bromo-3-fluorobenzenecarbonitrile, 3-cyano-5-fluorobromobenzene, 3-bromo 5-fluorobenzonitrile, 3-bromo-5-fluoro-benzonitrile, 3-bromo-5-fluorobenzenecarbonitrile, pubchem3220, acmc-209yem, 3-fluoro-5-bromobenzonitile | |
| FC1=CC(Br)=CC(=C1)C#N | |
| 200.01 | |
| 200.01 | |
| Crystalline Powder |
Safety and Handling
TSCA : No