missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-fluorobenzonitrile 98.0+%, TCI America™

Catalog No. B46001G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46001G 1 g
B46005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46001G Supplier TCI America Supplier No. B46001G

Chemical Identifiers

CAS 179898-34-1
Molecular Formula C7H3BrFN
Molecular Weight (g/mol) 200.01
MDL Number MFCD04038227
InChI Key IADLVSLZPQYXIF-UHFFFAOYSA-N
Synonym 3-fluoro-5-bromobenzonitrile, benzonitrile, 3-bromo-5-fluoro, 5-bromo-3-fluorobenzenecarbonitrile, 3-cyano-5-fluorobromobenzene, 3-bromo 5-fluorobenzonitrile, 3-bromo-5-fluoro-benzonitrile, 3-bromo-5-fluorobenzenecarbonitrile, pubchem3220, acmc-209yem, 3-fluoro-5-bromobenzonitile
PubChem CID 2783330
IUPAC Name 3-bromo-5-fluorobenzonitrile
SMILES FC1=CC(Br)=CC(=C1)C#N

Specifications

Melting Point 46°C
Color White
Quantity 1 g
UN Number 3439
Formula Weight 200.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-5-fluorobenzonitrile