missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-fluorobenzonitrile, 98%

CAS: 179898-34-1 | C7H3BrFN | 200.01 g/mol

$41.66 - $264.79

Chemical Identifiers

CAS 179898-34-1
Molecular Formula C7H3BrFN
Molecular Weight (g/mol) 200.01
MDL Number MFCD04038227
InChI Key IADLVSLZPQYXIF-UHFFFAOYSA-N
Synonym 3-fluoro-5-bromobenzonitrile, benzonitrile, 3-bromo-5-fluoro, 5-bromo-3-fluorobenzenecarbonitrile, 3-cyano-5-fluorobromobenzene, 3-bromo 5-fluorobenzonitrile, 3-bromo-5-fluoro-benzonitrile, 3-bromo-5-fluorobenzenecarbonitrile, pubchem3220, acmc-209yem, 3-fluoro-5-bromobenzonitile
PubChem CID 2783330
IUPAC Name 3-bromo-5-fluorobenzonitrile
SMILES FC1=CC(Br)=CC(=C1)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2606603
View Documents
Thermo Scientific Chemicals
H2606603
1 g
Each for $41.66
Only null left
 
AAH2606609
View Documents
Thermo Scientific Chemicals
H2606609
10 g
Each for $264.79
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

179898-34-1
200.01
IADLVSLZPQYXIF-UHFFFAOYSA-N
2783330
FC1=CC(Br)=CC(=C1)C#N
C7H3BrFN
MFCD04038227
3-fluoro-5-bromobenzonitrile, benzonitrile, 3-bromo-5-fluoro, 5-bromo-3-fluorobenzenecarbonitrile, 3-cyano-5-fluorobromobenzene, 3-bromo 5-fluorobenzonitrile, 3-bromo-5-fluoro-benzonitrile, 3-bromo-5-fluorobenzenecarbonitrile, pubchem3220, acmc-209yem, 3-fluoro-5-bromobenzonitile
3-bromo-5-fluorobenzonitrile

Specifications

179898-34-1
MFCD04038227
UN3439
IADLVSLZPQYXIF-UHFFFAOYSA-N
3-bromo-5-fluorobenzonitrile
2783330
98%
C7H3BrFN
1 g
3-fluoro-5-bromobenzonitrile, benzonitrile, 3-bromo-5-fluoro, 5-bromo-3-fluorobenzenecarbonitrile, 3-cyano-5-fluorobromobenzene, 3-bromo 5-fluorobenzonitrile, 3-bromo-5-fluoro-benzonitrile, 3-bromo-5-fluorobenzenecarbonitrile, pubchem3220, acmc-209yem, 3-fluoro-5-bromobenzonitile
FC1=CC(Br)=CC(=C1)C#N
200.01
200.01
3-Bromo-5-fluorobenzonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only