missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-fluorobenzonitrile, 98%

Catalog No. AAH2606603
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2606603 1 g
AAH2606609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2606603 Supplier Thermo Scientific Chemicals Supplier No. H2606603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 179898-34-1 | C7H3BrFN | 200.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 179898-34-1
Molecular Formula C7H3BrFN
Molecular Weight (g/mol) 200.01
MDL Number MFCD04038227
InChI Key IADLVSLZPQYXIF-UHFFFAOYSA-N
Synonym 3-fluoro-5-bromobenzonitrile, benzonitrile, 3-bromo-5-fluoro, 5-bromo-3-fluorobenzenecarbonitrile, 3-cyano-5-fluorobromobenzene, 3-bromo 5-fluorobenzonitrile, 3-bromo-5-fluoro-benzonitrile, 3-bromo-5-fluorobenzenecarbonitrile, pubchem3220, acmc-209yem, 3-fluoro-5-bromobenzonitile
PubChem CID 2783330
IUPAC Name 3-bromo-5-fluorobenzonitrile
SMILES FC1=CC(Br)=CC(=C1)C#N

Specifications

Quantity 1 g
UN Number UN3439
Formula Weight 200.01
Percent Purity 98%
Chemical Name or Material 3-Bromo-5-fluorobenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only