missing translation for 'onlineSavingsMsg'
Learn More

3'-Bromo-5'-chloro-2'-hydroxyacetophenone 97.0+%, TCI America™

Catalog No. p-7119571
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22365G 5 g
B223625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22365G Supplier TCI America Supplier No. B22365G

Chemical Identifiers

CAS 59443-15-1
Molecular Formula C8H6BrClO2
Molecular Weight (g/mol) 249.49
MDL Number MFCD00857068
InChI Key FFAVKFQPEOGJOA-UHFFFAOYSA-N
Synonym 2-Acetyl-6-bromo-4-chlorophenol
PubChem CID 2735546
IUPAC Name 1-(3-bromo-5-chloro-2-hydroxyphenyl)ethan-1-one
SMILES CC(=O)C1=CC(Cl)=CC(Br)=C1O

Specifications

Melting Point 100°C
Color Yellow
Quantity 5 g
Formula Weight 249.49
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Bromo-5′-chloro-2′-hydroxyacetophenone